CAS RN 447401-91-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 1-Ethylpyrazole-4-carboxylate, TRC
CAS: 447401-91-4 Molecular Formula: C8H12N2O2 Molecular Weight (g/mol): 168.19 Synonym: 1-Ethyl-1H-pyrazole-4-carboxylic Ecid Ethyl Ester,Ethyl 1-Ethyl-1H-pyrazole-4-carboxylate SMILES: CCOC(=O)c1cnn(CC)c1