CAS RN 4467-06-5
CAS RN 4467-06-5
IUPAC Name:
2-(4-methylphenyl)pyridine
Synonyms:
2-(4-methylphenyl)pyridine
Molecular Weight (g/mol):
169.23
Molecular Formula:
C12H11N
InChi Key:
KJNZQKYSNAQLEO-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CC=CC=N1
1
–
1
of
1
results
1
2-(p-Tolyl)pyridine, 98%
CAS: 4467-06-5 Molecular Formula: C12H11N Molecular Weight (g/mol): 169.23 MDL Number: MFCD00006281 InChI Key: KJNZQKYSNAQLEO-UHFFFAOYSA-N Synonym: 2-p-tolyl pyridine,2-4-methylphenyl pyridine,2-p-tolylpyridine,2-4-tolyl pyridine,pyridine, 2-4-methylphenyl,pyridine,2-p-tolyl,2-p-toluyl pyridine,pubchem16062,4-2-pyridyl toluene PubChem CID: 78219 IUPAC Name: 2-(4-methylphenyl)pyridine SMILES: CC1=CC=C(C=C1)C2=CC=CC=N2