accessibility menu, dialog, popup

UserName

CAS RN 444892-03-9

CH3H3CNHN

CAS RN 444892-03-9

IUPAC Name: 1-benzyl-3-(2-methylpropyl)piperazine
Synonyms: 1-benzyl-3-(2-methylpropyl)piperazine
Molecular Weight (g/mol): 232.37
Molecular Formula: C15H24N2
InChi Key: HQMROSCVRBNRRZ-UHFFFAOYNA-N
SMILES: CC(C)CC1CN(CC2=CC=CC=C2)CCN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results