accessibility menu, dialog, popup

UserName

H3C(R)OOPOOO

CAS RN 4378-40-9

IUPAC Name: (2R)-2-phosphonatobutanoate
Synonyms: (2R)-2-phosphonatobutanoate
Molecular Weight (g/mol): 165.06
Molecular Formula: C4H6O5P
InChi Key: PWSXRGRLZKVHLW-GSVOUGTGSA-K
SMILES: CC[C@H](C([O-])=O)P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results