accessibility menu, dialog, popup

UserName

H3COOHNFF

CAS RN 433967-28-3

IUPAC Name: 4-butoxy-N-(2,4-difluorophenyl)benzamide
Synonyms: 4-butoxy-N-(2,4-difluorophenyl)benzamide
Molecular Weight (g/mol): 305.33
Molecular Formula: C17H17F2NO2
InChi Key: AHCYOTLTLQTPSU-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C(=O)NC1=C(F)C=C(F)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results