accessibility menu, dialog, popup

UserName

BOOCH3HOOH

CAS RN 4334-87-6

IUPAC Name: [3-(ethoxycarbonyl)phenyl]boronic acid
Synonyms: 3-(ethoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 193.99
Molecular Formula: C9H11BO4
InChi Key: REHVCPNQQBDOJJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results