CAS RN 429658-95-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dabigatran (ethyl ester), MedChemExpress
MedChemExpress Dabigatran ethyl ester is an emerging oral anticoagulant which is a direct inhibitor of thrombin activity.