accessibility menu, dialog, popup

UserName

CAS RN 42965-55-9

IUPAC Name: 5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione; sulfuric acid
Synonyms: 5,6-diaminouracil; sulfuric acid
Molecular Weight (g/mol): 240.19
Molecular Formula: C4H8N4O6S
InChi Key: IKARJSDZQCSEJX-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results