accessibility menu, dialog, popup

UserName

H2N(S)BrHOO

CAS RN 42538-40-9

IUPAC Name: (2S)-2-amino-3-(2-bromophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2-bromophenyl)propanoic acid
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: JFVLNTLXEZDFHW-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=CC=C1Br)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)