CAS RN 42538-01-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Z-Ile-Ile, TRC
CAS: 42538-01-2 Molecular Formula: C20H30N2O5 Molecular Weight (g/mol): 378.47 Synonym: N-[N-[(Phenylmethoxy)carbonyl]-L-isoleucyl]-L-isoleucine SMILES: CC[C@H](C)[C@H](NC(=O)[C@@H](NC(=O)OCc1ccccc1)[C@@H](C)CC)C(=O)O