accessibility menu, dialog, popup

UserName

CAS RN 4214-79-3

ClNHO

CAS RN 4214-79-3

IUPAC Name: 5-chloro-1,2-dihydropyridin-2-one
Synonyms: 5-chloro-1H-pyridin-2-one
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: SZFUWUOHDRMCKD-UHFFFAOYSA-N
SMILES: ClC1=CNC(=O)C=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)