CAS RN 41944-01-8
IUPAC Name:
bismuth(3+) potassium tetraiodide
Synonyms:
bismuth(3+) potassium tetraiodide
Molecular Weight (g/mol):
755.7
Molecular Formula:
BiI4K
InChi Key:
ONBIUAZBGHXJDM-UHFFFAOYSA-J
SMILES:
[K+].[I-].[I-].[I-].[I-].[Bi+3]
1
–
4
of
4
results
2
Bismuth Potassium Iodide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bismuth Potassium Iodide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Bismuth Potassium Iodide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.