CAS RN 4151-33-1
CAS RN 4151-33-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyruvic Acid Potassium Salt, TRC
CAS: 4151-33-1 Molecular Formula: C3H3KO3 Molecular Weight (g/mol): 126.15 Synonym: Potassium Pyruvate;2-Oxopropanoic Acid Potassium Salt; IUPAC Name: potassium;2-oxopropanoate SMILES: [K+].CC(=O)C(=O)[O-]