accessibility menu, dialog, popup

UserName

CH3NHNSNH2

CAS RN 41440-07-7

IUPAC Name: (3-methylpyridin-2-yl)thiourea
Synonyms: 3-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: GHHXGWZMJBHPOM-UHFFFAOYSA-N
SMILES: CC1=C(NC(N)=S)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results