accessibility menu, dialog, popup

UserName

CH3NNH3COHSOOO

CAS RN 412009-61-1

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium hydrogen sulfate
Synonyms: 1-ethyl-3-methylimidazolium hydrogen sulfate
Molecular Weight (g/mol): 208.23
Molecular Formula: C6H12N2O4S
InChi Key: HZKDSQCZNUUQIF-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)