CAS RN 41078-02-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Enprofylline, TRC
CAS: 41078-02-8 Molecular Formula: C8H10N4O2 Molecular Weight (g/mol): 194.19 Synonym: 3,7-Dihydro-3-propyl-1H-purine-2,6-dione,3-Propylxanthine,D 4028,3,9-Dihydro-3-propyl-1H-purine-2,6-dione,3-propyl-1H-purine-2,6(3H,7H)-dione SMILES: CCCN1C(=O)NC(=O)c2nc[nH]c12