accessibility menu, dialog, popup

UserName

HO(R)NHO

CAS RN 40987-24-4

IUPAC Name: (2R)-4-benzyl-2-(hydroxymethyl)morpholin-4-ium
Synonyms: (2R)-4-benzyl-2-(hydroxymethyl)morpholin-4-ium
Molecular Weight (g/mol): 208.28
Molecular Formula: C12H18NO2
InChi Key: WQNIKIMRIXHNFF-GFCCVEGCSA-O
SMILES: OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)