accessibility menu, dialog, popup

UserName

OHClFFF

CAS RN 40889-91-6

IUPAC Name: 2-chloro-5-(trifluoromethyl)phenol
Synonyms: 2-chloro-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: SUIZARUATMVYRT-UHFFFAOYSA-N
SMILES: OC1=CC(=CC=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results