CAS RN 407582-49-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Febuxostat 67M-4, TRC
CAS: 407582-49-4 Molecular Formula: C16H14N2O5S Molecular Weight (g/mol): 346.36 Synonym: 2-[4-(2-Carboxypropoxy)-3-cyanophenyl]-4-methyl-5-thiazolecarboxylic Acid IUPAC Name: 2-[4-(2-carboxypropoxy)-3-cyanophenyl]-4-methyl-1,3-thiazole-5-carboxylic acid SMILES: O=C1N(C)C(=O)c2c(ncn2)N1