accessibility menu, dialog, popup

UserName

CH3OOHNI

CAS RN 40740-41-8

IUPAC Name: methyl 4-iodo-1H-pyrrole-2-carboxylate
Synonyms: methyl 4-iodo-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 251.02
Molecular Formula: C6H6INO2
InChi Key: KRNGDJYQOKCQCD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(I)=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results