accessibility menu, dialog, popup

UserName

CAS RN 404827-78-7

FNHNH2N

CAS RN 404827-78-7

IUPAC Name: 4-fluoro-1H-indazol-3-amine
Synonyms: 4-fluoro-1H-indazol-3-amine
Molecular Weight (g/mol): 151.14
Molecular Formula: C7H6FN3
InChi Key: HTMNYSASBRVPKD-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=CC(F)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results