CAS RN 4021-07-2
IUPAC Name:
3-methylpyridine-2-carboxylic acid
Synonyms:
3-methylpyridine-2-carboxylic acid
Molecular Weight (g/mol):
137.14
Molecular Formula:
C7H7NO2
InChi Key:
LMHIBYREWJHKNZ-UHFFFAOYSA-N
SMILES:
CC1=CC=CN=C1C(O)=O
1
–
1
of
1
results
1
3-Methylpicolinic acid, 98%, Thermo Scientific™
CAS: 4021-07-2 Molecular Formula: C7H7NO2 Molecular Weight (g/mol): 137.14 MDL Number: MFCD00191553 InChI Key: LMHIBYREWJHKNZ-UHFFFAOYSA-N IUPAC Name: 3-methylpyridine-2-carboxylic acid