accessibility menu, dialog, popup

UserName

CAS RN 400-54-4

CH3OOFFF

CAS RN 400-54-4

IUPAC Name: 1,1,1-trifluorohexane-2,4-dione
Synonyms: 1,1,1-trifluorohexane-2,4-dione
Molecular Weight (g/mol): 168.12
Molecular Formula: C6H7F3O2
InChi Key: SBRYTXXHTKCMFT-UHFFFAOYSA-N
SMILES: CCC(=O)CC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results