accessibility menu, dialog, popup

UserName

CAS RN 39895-55-1

CH3H3CCH3NH2

CAS RN 39895-55-1

IUPAC Name: 1-(4-tert-butylphenyl)methanamine
Synonyms: 1-(4-tert-butylphenyl)methanamine
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: MPWSRGAWRAYBJK-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CN)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)