accessibility menu, dialog, popup

UserName

BrOBr

CAS RN 3988-03-2

IUPAC Name: bis(4-bromophenyl)methanone
Synonyms: bis-(4-bromo-phenyl)-methanone
Molecular Weight (g/mol): 340.01
Molecular Formula: C13H8Br2O
InChi Key: LFABNOYDEODDFX-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results