accessibility menu, dialog, popup

UserName

CAS RN 3977-23-9

CH3CH3ONH2NHN

CAS RN 3977-23-9

IUPAC Name: 2-amino-5,6-dimethyl-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-5,6-dimethyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 139.16
Molecular Formula: C6H9N3O
InChi Key: APWRLAZEMYLHKZ-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)N=C(N)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results