CAS RN 3973-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Propynol Ethoxylate (>80%), TRC
CAS: 3973-18-0 Molecular Formula: C5H8O2 Molecular Weight (g/mol): 100.12 Synonym: 2-(2-Propyn-1-yloxy)ethanol,2-(2-Propynyloxy)ethanol,2-Propynoxyethanol,3-(2-Hydroxyethoxy)-1-propyne,4-Oxa-1-hexyn-6-ol,Ethylene Glycol Monopropargyl Ether IUPAC Name: 2-prop-2-ynoxyethanol SMILES: OCCOCC#C