accessibility menu, dialog, popup

UserName

BrN

CAS RN 3964-04-3

IUPAC Name: 4-bromoquinoline
Synonyms: 4-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: SUXIPCHEUMEUSV-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results