accessibility menu, dialog, popup

UserName

CAS RN 3939-13-7

FNN

CAS RN 3939-13-7

IUPAC Name: 2-fluoropyridine-3-carbonitrile
Synonyms: 2-fluoropyridine-3-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: USIDQCCXMGJOJM-UHFFFAOYSA-N
SMILES: FC1=NC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results