accessibility menu, dialog, popup

UserName

CH3NNH2N

CAS RN 39270-45-6

IUPAC Name: 2-(4-methyl-1,4-diazepan-1-yl)ethan-1-amine
Synonyms: 2-(4-methyl-1,4-diazepan-1-yl)ethanamine
Molecular Weight (g/mol): 157.26
Molecular Formula: C8H19N3
InChi Key: DDYKTHJBSBIWNR-UHFFFAOYSA-N
SMILES: CN1CCCN(CCN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results