accessibility menu, dialog, popup

UserName

BrNNNH2

CAS RN 392307-25-4

IUPAC Name: 4-(3-bromophenyl)pyrimidin-2-amine
Synonyms: 4-(3-bromophenyl)pyrimidin-2-amine
Molecular Weight (g/mol): 250.1
Molecular Formula: C10H8BrN3
InChi Key: LJDRXCOKWBGOOY-UHFFFAOYSA-N
SMILES: NC1=NC=CC(=N1)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results