CAS RN 3913-68-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2S,4S)-4-Hydroxy-L-glutamic Acid, TRC
CAS: 3913-68-6 Molecular Formula: C5H9NO5 Molecular Weight (g/mol): 163.13 Synonym: (4S)-4-Hydroxy-L-glutamic Acid,(2S,4S)-g-4-Hydroxy-L-glutamic Acid,L-threo-4-Hydroxyglutamic Acid,threo-4-Hydroxy-L-glutamic Acid,L-allo-4-Hydroxyglutamic acid,L-allo-gamma-Hydroxyglutamate,threo-gamma-Hydroxy-L-glutamic Acid,H-(2S,4S)-Glu(g-OH)-OH IUPAC Name: (2S,4S)-2-amino-4-hydroxypentanedioic acid SMILES: N[C@@H](C[C@H](O)C(=O)O)C(=O)O