accessibility menu, dialog, popup

UserName

CAS RN 3913-17-5

CH3CH3ClN

CAS RN 3913-17-5

IUPAC Name: 2-chloro-4,8-dimethylquinoline
Synonyms: 2-chloro-4,8-dimethylquinoline
Molecular Weight (g/mol): 191.66
Molecular Formula: C11H10ClN
InChi Key: UAKXMKQZURGJKF-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC2=C(C)C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results