accessibility menu, dialog, popup

UserName

CH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHRu(E/Z)(E/Z)(E/Z)

CAS RN 38625-54-6

IUPAC Name: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) ruthenium
Synonyms: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) ruthenium
Molecular Weight (g/mol): 653.91
Molecular Formula: C33H60O6Ru
InChi Key: TZPFVUZFVRIDGB-UHFFFAOYSA-N
SMILES: [Ru].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results