CAS RN 38557-82-3
CAS RN 38557-82-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Pyridinecarboxamide 1-Oxide, TRC
CAS: 38557-82-3 Molecular Formula: C6 H6 N2 O2 Molecular Weight (g/mol): 138.12 Synonym: 4-Pyridinecarboxamide, 1-oxide,Isonicotinamide, 1-oxide (6CI,7CI),1-Oxidopyridin-1-ium-4-carboxamide,Isonicotinamide N-oxide,NSC 18386,Isonicotinamide 1-oxide,4-Pyridinecarboxamide 1-oxide,4-Pyridinecarboxamide N-oxide IUPAC Name: 1-oxidopyridin-1-ium-4-carboxamide SMILES: NC(=O)c1cc[n+]([O-])cc1