CAS RN 38440-79-8
CAS RN 38440-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Deoxy-2-fluoro-D-mannose, TRC
CAS: 38440-79-8 Molecular Formula: C6 H11 F O5 Molecular Weight (g/mol): 182.15 Synonym: D-Mannose, 2-deoxy-2-fluoro-,2-Deoxy-2-fluoro-D-mannose IUPAC Name: (3S,4S,5S,6R)-3-fluoro-6-(hydroxymethyl)oxane-2,4,5-triol SMILES: OC[C@H]1OC(O)[C@@H](F)[C@@H](O)[C@@H]1O