accessibility menu, dialog, popup

UserName

ClClClClClCl

CAS RN 38380-07-3

IUPAC Name: 2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Synonyms: 2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: BTAGRXWGMYTPBY-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1Cl)C1=CC=C(Cl)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (2)
Concentration 
  • (2)
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)