accessibility menu, dialog, popup

UserName

OOOOOOOOOHoHo

CAS RN 38245-34-0

IUPAC Name: diholmium(3+) hydrate tricarbonate
Synonyms: diholmium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 527.9
Molecular Formula: C3H2Ho2O10
InChi Key: XXHLSAONJAUANQ-UHFFFAOYSA-H
SMILES: O.[Ho+3].[Ho+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results