CAS RN 38210-27-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Frutinone A, TRC
CAS: 38210-27-4 Molecular Formula: C16H8O4 Molecular Weight (g/mol): 264.23 Synonym: 6H,7H-[1]Benzopyrano[4,3-b][1]benzopyran-6,7-dione SMILES: O=C1Oc2ccccc2C3=C1C(=O)c4ccccc4O3