accessibility menu, dialog, popup

UserName

CAS RN 37839-81-9

N(R)N(R)O(S)(R)NNOHNH2OPOOONa

CAS RN 37839-81-9

IUPAC Name: sodium 6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate hydrate
Synonyms: sodium 6-(6-aminopurin-9-yl)-7-hydroxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate hydrate
Molecular Weight (g/mol): 369.21
Molecular Formula: C10H13N5NaO7P
InChi Key: FKBJSPWBAQGFIV-UHFFFAOYNA-M
SMILES: O.[Na+].NC1=C2N=CN(C3OC4COP([O-])(=O)OC4C3O)C2=NC=N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)