CAS RN 37836-90-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MPO-IN-28, MedChemExpress
MedChemExpress MPO-IN-28 (Compound 28) is a myeloperoxidase (MPO) inhibitor with an IC50 of 44 nM.