accessibility menu, dialog, popup

UserName

CAS RN 3777-70-6

H3CO

CAS RN 3777-70-6

IUPAC Name: 2-hexylfuran
Synonyms: 2-hexylfuran
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: XBLCAKKYMZVLPU-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results