CAS RN 3749-51-7
IUPAC Name:
2-hydroxy-6-methyl-1,4-dihydropyridin-4-one
Synonyms:
2-hydroxy-6-methyl-1H-pyridin-4-one
Molecular Weight (g/mol):
125.13
Molecular Formula:
C6H7NO2
InChi Key:
WKGSLYHMRQRARV-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)C=C(O)N1
1
–
1
of
1
results
1
2,4-dihydroxy-6-methylpyridine, 97%, Thermo Scientific Chemicals
CAS: 3749-51-7 Molecular Formula: C6H7NO2 Molecular Weight (g/mol): 125.13 MDL Number: MFCD01860849 InChI Key: WKGSLYHMRQRARV-UHFFFAOYSA-N Synonym: 6-methylpyridine-2,4-diol,2,4-dihydroxy-6-methylpyridine,4-hydroxy-6-methylpyridin-2 1h-one,4-hydroxy-6-methyl-2 1h-pyridinone,2 1h-pyridinone, 4-hydroxy-6-methyl,6-methyl-2,4-pyridinediol,2-hydroxy-6-methyl-1h-pyridin-4-one,2,4-dihydroxy-6-methyl-pyridine,4-hydroxy-6-methyl-2-pyridinone,2 1h-pyridone, 4-hydroxy-6-methyl PubChem CID: 54691419 IUPAC Name: 4-hydroxy-6-methyl-1H-pyridin-2-one SMILES: CC1=CC(=CC(=O)N1)O