CAS RN 3729-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,5-Diphenyl-3-methylpyrazole, TRC
CAS: 3729-90-6 Molecular Formula: C16H14N2 Molecular Weight (g/mol): 234.3 SMILES: Cc1cc(c2ccccc2)n(n1)c3ccccc3