accessibility menu, dialog, popup

UserName

CAS RN 37264-20-3

OSiCr

CAS RN 37264-20-3

IUPAC Name: oxidaniumylidynesilanide chromium
Synonyms: silicon monoxide chromium
Molecular Weight (g/mol): 96.08
Molecular Formula: CrOSi
InChi Key: ASAMIKIYIFIKFS-UHFFFAOYSA-N
SMILES: [Cr].[O+]#[Si-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results