CAS RN 3720-97-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,5-Dihydroxytetrahydro-2H-imidazol-2-one, TRC
CAS: 3720-97-6 Molecular Formula: C3H6N2O3 Molecular Weight (g/mol): 118.09 SMILES: OC1NC(=O)NC1O