CAS RN 36953-42-1
IUPAC Name:
3-bromo-4-chloropyridine
Synonyms:
3-bromo-4-chloropyridine
Molecular Weight (g/mol):
192.44
Molecular Formula:
C5H3BrClN
InChi Key:
QADXKWUCCGPQNR-UHFFFAOYSA-N
SMILES:
ClC1=CC=NC=C1Br
1
–
1
of
1
results
1
3-Bromo-4-chloropyridine, 98%
CAS: 36953-42-1 Molecular Formula: C5H3BrClN Molecular Weight (g/mol): 192.44 MDL Number: MFCD00234008 InChI Key: QADXKWUCCGPQNR-UHFFFAOYSA-N Synonym: pyridine, 3-bromo-4-chloro,3-bromo-4-chloro-pyridine,4-chloro-3-bromopyridine,acmc-1ai1d,3-bromo-4-chloropyridine,,ksc222g5j,pyridine,3-bromo-4-chloro-,hydrochloride 1:1 PubChem CID: 817696 IUPAC Name: 3-bromo-4-chloropyridine SMILES: C1=CN=CC(=C1Cl)Br