CAS RN 361436-29-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Di-(tert-Butyloxycarbonyl) Serotonin, TRC
CAS: 361436-29-5 Molecular Formula: C20H28N2O5 Molecular Weight (g/mol): 376.45 Synonym: 3-[2-[[(1,1-Dimethylethoxy)carbonyl]amino]ethyl]-5-hydroxy-1H-indole-1-carboxylic Acid 1,1-Dimethylethyl Ester SMILES: CC(C)(C)OC(=O)NCCc1cn(C(=O)OC(C)(C)C)c2ccc(O)cc12