CAS RN 3604-87-3
CAS RN 3604-87-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Alpha-Ecdysone ( >90%), TRC
CAS: 3604-87-3 Molecular Formula: C27 H44 O6 Molecular Weight (g/mol): 464.63 Synonym: (2β,3β,5β,22R)-2,3,14,22,25-Pentahydroxycholest-7-en-6-one IUPAC Name: (2S,3R,5R,9R,10R,13R,14S,17R)-17-[(1S,2R)-2,5-dihydroxy-1,5-dimethyl-hexyl]-2,3,14-trihydroxy-10,13-dimethyl-2,3,4,5,9,11,12,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-6-one SMILES: C[C@H]([C@H](O)CCC(C)(C)O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C