CAS RN 359586-64-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S,2S)-Fmoc-acpc, TRC
CAS: 359586-64-4 Molecular Formula: C21H21NO4 Molecular Weight (g/mol): 351.4 Synonym: Fmoc-(1S,2S)-2-aminocyclopentane carboxylic Acid SMILES: OC(=O)[C@H]1CCC[C@@H]1NC(=O)OCC2c3ccccc3c4ccccc24